Skip to Content
Merck
All Photos(1)

Key Documents

324R-1

Sigma-Aldrich

PSA (EP109) Rabbit Monoclonal Primary Antibody

Synonym(s):

APS, KLK2A1, P-30 antigen, gamma-seminoprotein, hK3, kallikrein-3, kallikrein-related peptidase 3, prostate specific antigen, semenogelase, seminin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

EP109, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (324R-14)
vial of 0.5 mL concentrate (324R-15)
vial of 1.0 mL concentrate (324R-16)
vial of 1.0 mL pre-dilute ready-to-use (324R-17)
vial of 7.0 mL pre-dilute ready-to-use (324R-18)

manufacturer/tradename

Cell Marque

concentration

0.1-0.5 μg/mL (predilute)
10-500 μg/mL (concentrate)

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500 (concentrated)

isotype

IgG

control

prostate carcinoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... KLK3(354)

General description

Prostate-Specific Antigen (PSA) is a 33 kDa protein primarily produced by the prostatic epithelium and the epithelial lining of the periurethral glands. PSA is strongly expressed in both normal and neoplastic prostatic tissue. Anti-PSA is most useful in determining the prostatic origin of carcinomas in non-prostate tissues (metastatic disease) using IHC techniques.,,,

Quality

Canada - IVD
European Union - IVD
Japan - RUO
United States - IVD

Caution

Do not freeze

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prostate specific antigen: a decade of discovery -- what we have learned and where we are going.
Polascik, T.J., et al.
The Journal of Urology, 162, 293-306 (1999)
Immunohistochemical labelling for prostate specific antigen in nonprostatic tissues.
Alanen, K.A., et al.
Pathology Research and Practice, 192, 233-237 (1996)
Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literature.
Varma, M., and Jasani, B.
Histopathology, 47, 1-16 (2005)
Prostate-specific antigen.
Stenman, U.H., et al.
Seminars in Cancer Biology, 9, 83-93 (1999)
M Varma et al.
Histopathology, 47(1), 1-16 (2005-06-29)
Varma M & Jasani B (2005) Histopathology47, 1-16 Diagnostic utility of immunohistochemistry in morphologically difficult prostate cancer: review of current literatureImmunohistochemistry is widely used to distinguish prostate cancer from benign mimics and to establish the prostatic origin of poorly differentiated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service