Merck
  • Osteo-odonto-keratoprosthesis (OOKP) and the testing of three different adhesives for bonding bovine teeth with optical poly-(methyl methacrylate) (PMMA) cylinder.
Osteo-odonto-keratoprosthesis (OOKP) and the testing of three different adhesives for bonding bovine teeth with optical poly-(methyl methacrylate) (PMMA) cylinder.
The British journal of ophthalmology (2013-07-16)
K Weisshuhn, I Berg, D Tinner, C Kunz, M M Bornstein, M Steineck, K Hille, D Goldblum
ABSTRACT
Preparation of the lamina during osteo-odonto-keratoprosthesis (OOKP) design is complex, and its longevity and watertightness important. To date, only acrylic bone cements have been used for bonding the optical cylinder to the tooth dentine. Our aim was to evaluate different dental adhesives for OOKP preparation. Specimens of bovine teeth were produced by preparing 1.5-mm thick dentine slices with holes having a diameter of 3.5 mm. Each group (n=10 per group) was luted with either classic poly-(methyl methacrylate) (PMMA) bone cement, universal resin cement or glass ionomer cement. All specimens underwent force measurement using a uniaxial traction machine. The highest mean force required to break the bond was measured for PMMA bone cement (128.2 N) followed by universal resin cement (127.9 N), with no statistically significant difference. Glass ionomer cement showed significantly lower force resistance (78.1 N). Excellent bonding strength combined with easy application was found for universal resin cement, and thus, it is a potential alternative to acrylic bone cement in OOKP preparation.
Sigma-Aldrich
Poly(methyl methacrylate), CP
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic