Skip to Content
Merck
All Photos(1)

Documents

81497

Supelco

Poly(methyl methacrylate)

analytical standard, for GPC, 10,000

Synonym(s):

PMMA, Poly(methacrylic acid methyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2C(CH3)(CO2CH3)]n
CAS Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

autoignition temp.

580 °F

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~10400
Mp ~10900
Mw ~10900

technique(s)

gel permeation chromatography (GPC): suitable

solubility

H2O: insoluble

density

1.200 g/cm3

Mw/Mn

~1.05

format

neat

storage temp.

2-8°C

InChI

1S/C5H9O2/c1-4(2)5(6)7-3/h1-3H3

InChI key

PMAMJWJDBDSDHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

François Therriault-Proulx et al.
Physics in medicine and biology, 58(7), 2073-2084 (2013-03-09)
The goal of this study was to evaluate the nature of the stem effect light produced within an optical fiber, to quantify its composition, and to evaluate the efficiency of the chromatic technique to remove the stem effect. Spectrometry studies
Taher Alizadeh et al.
Journal of hazardous materials, 248-249, 401-406 (2013-02-19)
It was demonstrated that a thin film of chemically exfoliated graphene flakes, blended with poly(methyl methacrylate), could be used as a chemiresistor sensor for the sensitive and selective determination of formaldehyde vapor. Formaldehyde adsorption on the sensing film led to
A K Pathak et al.
The Journal of clinical pediatric dentistry, 37(3), 335-339 (2013-07-17)
Biofilms on removable orthodontic appliances act as reservoir of microorganisms, capable of modifying the environmental condition of oral cavity and are difficult to be removed with routine hygiene measures. The present investigation includes enumeration, identification and numerical analysis of different
Clément M L Werner et al.
The Journal of bone and joint surgery. American volume, 95(7), 577-584 (2013-04-05)
In the treatment of vertebral compression fractures, vertebral body stenting with an expandable scaffold inserted before application of the bone cement was developed to impede secondary loss of vertebral height encountered in patients treated with balloon kyphoplasty. The purpose of
Sean M Barber et al.
Journal of neurosurgery. Spine, 18(5), 524-528 (2013-03-26)
Percutaneous sacroplasty is a procedure adapted from vertebroplasty, which is designed to ameliorate the painful morbidity associated with sacral insufficiency fractures without the invasiveness of open surgery. Early estimates of efficacy, according to several case reports and small series, appear

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service