Merck
  • Evaluation of solid adsorbent materials for cryogen-free trapping-gas chromatographic analysis of atmospheric C2-C6 non-methane hydrocarbons.
Evaluation of solid adsorbent materials for cryogen-free trapping-gas chromatographic analysis of atmospheric C2-C6 non-methane hydrocarbons.
Journal of chromatography. A (2006-10-03)
Jan Pollmann, Detlev Helmig, Jacques Hueber, David Tanner, Pieter P Tans
ABSTRACT
Nine commercial solid adsorbent materials (in order of decreasing surface area: Carboxen 1000, Carbosieve S III, molecular sieve 5A, molecular sieve 4A, silica gel, Carboxen 563, activated alumina, Carbotrap and Carboxen 1016) were investigated for their ability to trap and release C2-C6 non-methane hydrocarbons (NMHCs) in atmospheric samples for subsequent thermal desorption gas chromatography-flame ionization detection analysis (GC-FID). Recovery rates for 23 NMHCs and methyl chloride (CH3Cl) were determined. A microtrap filled with the three adsorbents Carbosieve S III, Carboxen 563 and Carboxen 1016 was found to allow for the analysis of the widest range of target analytes. A detection limit of approximately 3pptC [parts per trillion (carbon)] in a 1l air sample and a linear response over a wide range of volatilities and sample volumes was determined for this configuration. Water vapor in the sample air was found to causes interference in trapping and subsequent chromatographic analysis of light NMHCs. A Peltier-cooled, regenerable water trap inserted into the sample flow path was found to mitigate these problems and to allow quantitative and reproducible results for all analytes at all tested humidity conditions.
Product Description
Supelco
Graphsphere Adsorbent, matrix Graphsphere 2016, 60-80 mesh, bottle of 10 g
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
ORBO 101 Carbotrap® B (20/40), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 70 mm, pkg of 25 ea
Supelco
ORBO 100 on Carbotrap® B (20/40), 350/175 mg, W,W,W separators, O.D. × L 7 mm × 110 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 202, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg