Skip to Content
Merck
All Photos(2)

Key Documents

28274-U

Supelco

Carbosieve SIII Thermal Desorption Tube

greener alternative

stainless steel, O.D. x I.D. x L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea

Synonym(s):

Carbosieve S-III SS TD TUBE, 7 IN., FOR GERSTEL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Product Name

Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea

material

stainless steel TD tube

Quality Level

Agency

EPA TO-2

description

Sealed with TDS³ Storage Container

product line

Carbosieve

feature

preconditioned

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

active air sampling: suitable

O.D. × I.D. × L

6 mm × 4 mm × 7 in.

matrix

Carbosieve S-III carbon molecular sieve (CMS)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Gerstel® TDS A & TDS 2

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

Carbosieve SIII is a prepacked stainless steel sampling tube, fully compatible with the Gerstel TDS and TDSA instruments. It is thermally conditioned and batch tested for backpressure and background.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

GERSTEL is a registered trademark of Gerstel GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yannick Juillet et al.
The Analyst, 130(6), 977-982 (2005-05-25)
Among the chemicals belonging to the schedules of the Chemical Weapons Convention (CWC), sampling and analysis of highly volatile compounds such as hydrogen cyanide (HCN) require special consideration. The latter is present in numerous old chemical weapons that are stockpiled
C Y Peng et al.
Journal of environmental monitoring : JEM, 2(4), 313-324 (2001-03-16)
Air sampling, using sorbents, thermal desorption and gas chromatography, is a versatile method for identifying and quantifying trace levels of volatile organic compounds (VOCs). Thermal desorption can provide high sensitivity, appropropriate choices of sorbents and method parameters can accommodate a
R Magnusson et al.
Journal of chromatography. A, 1375, 17-26 (2014-12-17)
Phosphine (PH3), sulfuryl fluoride (SO2F2) and methyl bromide (CH3Br) are highly toxic chemical substances commonly used for fumigation, i.e., pest control with gaseous pesticides. Residues of fumigation agents constitute a health risk for workers affected, and therefore accurate methods for
R A Borders et al.
American Industrial Hygiene Association journal, 47(3), 158-163 (1986-03-01)
Thermal desorption is a more sensitive alternative to solvent desorption for the determination of acrylonitrile in air. A dual-bed collection tube (Tenax GC and Carbosieve B) was developed for collecting and concentrating low levels of acrylonitrile. Two thermal desorption techniques
Mohammad Reza Samarghandi et al.
Journal of research in health sciences, 14(3), 227-232 (2014-09-12)
Toluene is a volatile organic compound, one of 189 hazardous air pollutants (HAPs) and the most important pollutant found in most industries and indoor environments; owing to its adverse health, toluene must be treated before being released into the environment.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service