Merck
  • Management of spasticity in progressive multiple sclerosis: efficacy of repeated intrathecal triamcinolone acetonide administration.
Management of spasticity in progressive multiple sclerosis: efficacy of repeated intrathecal triamcinolone acetonide administration.
Current pharmaceutical design (2012-05-23)
Mazen Abu-Mugheisib, Reiner Benecke, Uwe K Zettl
ABSTRACT
Multiple sclerosis (MS) as a chronic disease presents a vast variability of neurological symptoms. Spasticity is one of the most common symptoms. The principle indication to treat spasticity is disability or handicap of capabilities. All pharmacological methods are attributed to a basic physical management. Different effective treatment schemes have been established. Nevertheless the value of intrathecal triamcinolone acetonid (TCA) in MS is still a controversial issue. Bearing in mind that only limited data are available and due to its invasive application form repeated TCA administration can be recommended as one therapy option in MS with a progressive clinical course and predominantly spinal symptoms.
Product Description
Supelco
Triamcinolone acetonide, analytical standard
Triamcinolone acetonide, European Pharmacopoeia (EP) Reference Standard
Triamcinolone acetonide for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Triamcinolone acetonide, United States Pharmacopeia (USP) Reference Standard