Skip to Content
Merck
All Photos(1)

Key Documents

1677002

USP

Triamcinolone acetonide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9α-Fluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide, 9α-Fluoro-16α-hydroxyprednisolone 16α,17α-acetonide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

triamcinolone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H]5OC(C)(C)O[C@@]45C(=O)CO

InChI

1S/C24H31FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-9,15-17,19,26,28H,5-6,10-12H2,1-4H3/t15-,16-,17-,19+,21-,22-,23-,24+/m0/s1

InChI key

YNDXUCZADRHECN-JNQJZLCISA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Triamcinolone acetonide USP reference standard intended for use in specified quality tests and assays.

Also used to prepare system suitability, standard, and standard stock solution for assay, impurity analysis, and performance tests according to the given below monographs of United States Pharmacopeia (USP):
  • Triamcinolone Hexacetonide
  • Triamcinolone Acetonide Injectable Suspension
  • Triamcinolone Acetonide Nasal Spray

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Triamcinolone Hexacetonide
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 36(5), 4492-4492 (2020)
Triamcinolone Acetonide Nasal Spray
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 46(5) (2021)
Triamcinolone Acetonide Injectable Suspension
United States Pharmacopeia and National Formulary
United States Pharmacopeia, 44(3), 4490-4490 (2020)
C M Jermak et al.
Survey of ophthalmology, 52(5), 503-522 (2007-08-28)
Triamcinolone acetonide has been effectively used in ocular therapeutics for over 50 years. Its use has increased dramatically in recent years for periocular and intraocular treatment of retinal vasculature disease and uveitis. This comprehensive review discusses the pharmacokinetics of triamcinolone
Kent W Small et al.
Ophthalmology, 121(4), 952-958 (2014-02-11)
To report a series of cases with fungal endophthalmitis occurring after intravitreal injection of triamcinolone derived from a single lot prepared by a compounding pharmacy. Retrospective, observational case series. Seventeen eyes treated with triamcinolone obtained from a single lot subsequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service