Merck
  • Multidisciplinary utilization of dimethyl sulfoxide: pharmacological, cellular, and molecular aspects.
Multidisciplinary utilization of dimethyl sulfoxide: pharmacological, cellular, and molecular aspects.
Biochemical pharmacology (2003-03-29)
Nuno C Santos, J Figueira-Coelho, J Martins-Silva, Carlota Saldanha
ABSTRACT
DMSO is an amphipathic molecule with a highly polar domain and two apolar methyl groups, making it soluble in both aqueous and organic media. It is one of the most common solvents for the in vivo administration of several water-insoluble substances. Despite being frequently used as a solvent in biological studies and as a vehicle for drug therapy, the side-effects of DMSO (undesirable for these purposes) are apparent from its utilization in the laboratory (both in vivo and in vitro) and in clinical settings. DMSO is a hydrogen-bound disrupter, cell-differentiating agent, hydroxyl radical scavenger, intercellular electrical uncoupler, intracellular low-density lipoprotein-derived cholesterol mobilizing agent, cryoprotectant, solubilizing agent used in sample preparation for electron microscopy, antidote to the extravasation of vesicant anticancer agents, and topical analgesic. Additionally, it is used in the treatment of brain edema, amyloidosis, interstitial cystitis, and schizophrenia. Several systemic side-effects from the use of DMSO have been reported, namely nausea, vomiting, diarrhea, hemolysis, rashes, renal failure, hypertension, bradycardia, heart block, pulmonary edema, cardiac arrest, and bronchospasm. Looking at the multitude of effects of DMSO brought to light by these studies, it is easily understood how many researchers working with DMSO (or studying one of its specific effects) might not be fully aware of the experiences of other groups who are working with it but in a different context.
Product Description
Supelco
Dimethyl Sulfoxide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.0%, suitable for absorption spectrum analysis
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, SAJ first grade, ≥99.0%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, JIS special grade, ≥99.0%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%