Merck
  • [The intensification of biological nitrogen fixation by using ammonium carbonate compounds and the routes of their action on microorganisms and plants].
[The intensification of biological nitrogen fixation by using ammonium carbonate compounds and the routes of their action on microorganisms and plants].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2000-08-10)
N N Mal'tseva, G I Vilesov, O E Davydova
ABSTRACT
The intensification of activity of associative and symbiotic nitrogen fixation while using low amounts of ammonium-carbonate compounds, carbonic-ammonium salts, in particular, has been established. The main ways and mechanisms of the action of ammonium-carbonate salts as a new and perspective agrochemical on physiological and biochemical processes of microorganisms and agricultural plants were presented on the basis of the analysis of literary information and the author's investigations.
Sigma-Aldrich
Ammonium carbonate, SAJ first grade, ≥20.0% NH3 basis
Sigma-Aldrich
Ammonium carbonate, ACS reagent, ≥30.0% NH3 basis
Sigma-Aldrich
Ammonium carbonate, puriss., meets analytical specification of NF, Ph. Franc., FCC
Sigma-Aldrich
Ammonium carbonate, 99.999% trace metals basis