Skip to Content
Merck
All Photos(1)

Key Documents

11204

Sigma-Aldrich

Ammonium carbonate

puriss., meets analytical specification of NF, Ph. Franc., FCC

Synonym(s):

Hartshorn salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2CO3
CAS Number:
Molecular Weight:
96.09
Beilstein:
4253487
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38020208
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

vapor density

2.7 (vs air)

Quality Level

grade

puriss.

form

powder or crystals

quality

meets analytical specification of NF, Ph. Franc., FCC

concentration

30.0-34.0% (NH3 , titration)

impurities

residual solvents, complies
≤0.0005% heavy metals (as Pb)
≤0.05% non-volatile matter

ign. residue

≤0.05% (as SO4)

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤3 mg/kg
Fe: ≤5 mg/kg
Pb: ≤3 mg/kg

suitability

complies for appearance of solution

SMILES string

N.N.OC(O)=O

InChI

1S/CH2O3.2H3N/c2-1(3)4;;/h(H2,2,3,4);2*1H3

InChI key

PRKQVKDSMLBJBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium carbonate is an ammonium salt. It is colorless and is composed of water soluble cubic crystals. It can be synthesized by reacting carbon dioxide with ammonia. It is widely employed as a CO2 source in fire extinguishers. Calorimetric estimation of enthalpies of absorption of CO2 in aqueous solutions of ammonium carbonate in the temperature range of 35-80°C have been reported.

Application

Ammonium carbonate may be employed in the preparation of 3,4-dihydropyrimidin-2(1H)-one and 1,4-dihydropyridine derivatives, via Biginelli and Hantzsch reactions. It can also be actively used as buffer in the separation, detection, and identification of the peptide fragments generated in the tryptic digests of ovalbumin and serum albumin, via size exclusion liquid chromatography and reverse phase liquid chromatography coupled to mass spectrometry.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Two-Dimensional SEC/RPLC Coupled to Mass Spectrometry for the Analysis of Peptides?
Opiteck.JG and Jorgenson.WJ
Journal of Analytical Chemistry, 69, 2283-2291 (1997)
Eagleson M.
Concise Encyclopedia Chemistry (1994)
Synthesis of 3, 4-dihydropyrimidin-2 (1H)-ones and 1, 4-dihydropyridines using ammonium carbonate in water.
Tamaddon F, et al.
Tetrahedron Letters, 51(8), 1187-1189 (2010)
A Polettini et al.
Journal of chromatography. B, Biomedical sciences and applications, 754(2), 447-459 (2001-05-08)
For the first time, an LC-MS-MS method has been developed for the simultaneous analysis of buprenorphine (BUP), norbuprenorphine (NBUP), and buprenorphine-glucuronide (BUPG) in plasma. Analytes were isolated from plasma by C18 SPE and separated by gradient RP-LC. Electrospray ionization and
Heat of absorption of CO2 in aqueous ammonia and ammonium carbonate/carbamate solutions.
Qin F, et al.
International Journal of Greenhouse Gas Control, 5(3), 405-412 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service