Merck
Dental caries and enamelin haplotype.
Journal of dental research (2014-02-04)
C Chaussain, N Bouazza, B Gasse, A G Laffont, S Opsahl Vital, T Davit-Béal, E Moulis, O Chabadel, M Hennequin, F Courson, D Droz, F Vaysse, O Laboux, H Tassery, J C Carel, A Alcais, J M Treluyer, C Beldjord, J Y Sire
ABSTRACT
In the literature, the enamelin gene ENAM has been repeatedly designated as a possible candidate for caries susceptibility. Here, we checked whether ENAM variants could increase caries susceptibility. To this aim, we sequenced coding exons and exon-intron boundaries of ENAM in 250 children with a severe caries phenotype and in 149 caries-free patients from 9 French hospital groups. In total, 23 single-nucleotide polymorphisms (SNPs) were found, but none appeared to be responsible for a direct change of ENAM function. Six SNPs had a high minor allele frequency (MAF) and 6 others were identified for the first time. Statistical and evolutionary analyses showed that none of these SNPs was associated with caries susceptibility or caries protection when studied separately and challenged with environmental factors. However, haplotype interaction analysis showed that the presence, in a same variant, of 2 exonic SNPs (rs7671281 and rs3796704; MAF 0.12 and 0.10, respectively), both changing an amino acid in the protein region encoded by exon 10 (p.I648T and p.R763Q, respectively), increased caries susceptibility 2.66-fold independent of the environmental risk factors. These findings support ENAM as a gene candidate for caries susceptibility in the studied population.
Product Description
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
L-Threonine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Isoleucine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Supelco
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
Arginine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Isoleucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
SAFC
L-Isoleucine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Arginine
Sigma-Aldrich
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Arginine, reagent grade, ≥98%
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginine, 99%, FCC, FG
Sigma-Aldrich
L-Arginine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture