Merck
  • Pro-TRH and pro-CRF expression in paraventricular nucleus of small litter-reared fasted adult rats.
Pro-TRH and pro-CRF expression in paraventricular nucleus of small litter-reared fasted adult rats.
The Journal of endocrinology (2014-01-28)
F Aréchiga-Ceballos, E Alvarez-Salas, G Matamoros-Trejo, M I Amaya, C García-Luna, P de Gortari
ABSTRACT
Neuroendocrine axes adapt to nutrient availability. During fasting, the function of the hypothalamus-pituitary-thyroid axis (HPT) is reduced, whereas that of the hypothalamus-pituitary-adrenal axis (HPA) is increased. Overfeeding-induced hyperleptinemia during lactation may alter the regulatory set point of neuroendocrine axes and their adaptability to fasting in adulthood. Hyperleptinemia is developed in rodents by litter size reduction during lactation; adult rats from small litters become overweight, but their paraventricular nucleus (PVN) TRH synthesis is unchanged. It is unclear whether peptide expression still responds to nutrient availability. PVN corticotropin-releasing factor (CRF) expression has not been evaluated in this model. We analyzed adaptability of HPT and HPA axes to fasting-induced low leptin levels of reduced-litter adult rats. Offspring litters were reduced to 2-3/dam (early-overfed) or maintained at 8/dam (controls, C). At 10 weeks old, a subset of animals from each group was fasted for 48 h and leptin, corticosterone, and thyroid hormones serum levels were analyzed. In brain, expressions of leptin receptor, NPY and SOCS3, were evaluated in arcuate nucleus, and those of proTRH and proCRF in PVN by real-time PCR. ProTRH expression in anterior and medial PVN subcompartments was assayed by in situ hybridization. Early-overfed adults developed hyperphagia and excessive weight, together with decreased proTRH expression in anterior PVN, supporting the anorexigenic effects of TRH. Early-overfed rats presented low PVN proTRH synthesis, whereas fasting did not induce a further reduction. Fasting-induced stress was unable to increase corticosterone levels, contributing to reduced body weight loss in early-overfed rats. We concluded that early overfeeding impaired the adaptability of HPT and HPA axes to excess weight and fasting in adults.
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)
Sigma-Aldrich
L-Pyroglutamic acid, BioXtra
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Sigma-Aldrich
L-Pyroglutamic acid, ≥99.0% (T)
Sigma-Aldrich
Thyrotropin releasing hormone, ≥98% (HPLC), powder
Supelco
Corticosterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pidolic acid, European Pharmacopoeia (EP) Reference Standard