Merck
  • Male-produced aggregation pheromone of the american palm weevil,Rhynchophorus palmarum (L.) (Coleoptera, Curculionidae): Collection, identification, electrophysiogical activity, and laboratory bioassay.
Male-produced aggregation pheromone of the american palm weevil,Rhynchophorus palmarum (L.) (Coleoptera, Curculionidae): Collection, identification, electrophysiogical activity, and laboratory bioassay.
Journal of chemical ecology (1991-11-01)
D Rochat, C Malosse, M Lettere, P H Ducrot, P Zagatti, M Renou, C Descoins
ABSTRACT
Male American palm weevils (APWs),Rhynchophorus palmarum (L.) produced two sex-specific compounds, which were disclosed by volatile collections on Supelpak-2 and gas chromatography. One was a minor compound, not always detected. The major male-produced volatile was identified as (2E)-6-methyl-2-hepten-4-ol through coupled gas chromatography-mass spectrometry and gas chromatography-Fourier transform infrared spectrometry, proton nuclear magnetic resonance spectrometry, and rational synthesis. We propose the trivial name rhynchophorol for this new molecule, which proved to be the essential component of the APW aggregation pheromone by electroantennography, coupled gas chromatography-electroantennography and behavioral bioassays.
Product Description
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
Supelpak-2SVM, pkg of 500 g
Supelco
Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 1000 g
Supelco
Supelpak-2, matrix styrene-divinylbenzene, 20-60 mesh, jar of 100 g
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea