Merck
  • Studies on the regulation of the human E1 subunit of the 2-oxoglutarate dehydrogenase complex, including the identification of a novel calcium-binding site.
Studies on the regulation of the human E1 subunit of the 2-oxoglutarate dehydrogenase complex, including the identification of a novel calcium-binding site.
The Biochemical journal (2014-02-06)
Craig T Armstrong, J L Ross Anderson, Richard M Denton
ABSTRACT
The regulation of the 2-oxoglutarate dehydrogenase complex is central to intramitochondrial energy metabolism. In the present study, the active full-length E1 subunit of the human complex has been expressed and shown to be regulated by Ca2+, adenine nucleotides and NADH, with NADH exerting a major influence on the K0.5 value for Ca2+. We investigated two potential Ca2+-binding sites on E1, which we term site 1 (D114ADLD) and site 2 (E139SDLD). Comparison of sequences from vertebrates with those from Ca2+-insensitive non-vertebrate complexes suggest that site 1 may be the more important. Consistent with this view, a mutated form of E1, D114A, shows a 6-fold decrease in sensitivity for Ca2+, whereas variant ∆site1 (in which the sequence of site 1 is replaced by A114AALA) exhibits an almost complete loss of Ca2+ activation. Variant ∆site2 (in which the sequence is replaced with A139SALA) shows no measurable change in Ca2+ sensitivity. We conclude that site 1, but not site 2, forms part of a regulatory Ca2+-binding site, which is distinct from other previously described Ca2+-binding sites.
Sigma-Aldrich
α-Ketoglutaric acid potassium salt, ≥98% (enzymatic)
Sigma-Aldrich
α-Ketoglutaric acid, 99.0-101.0% (T)
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, BioUltra
Sigma-Aldrich
α-Ketoglutaric acid sodium salt, ≥98% (titration)
Sigma-Aldrich
α-Ketoglutarate Dehydrogenase from porcine heart, buffered aqueous glycerol solution, 0.1-1.0 units/mg protein (Lowry)
Supelco
α-Ketoglutaric acid disodium salt hydrate, ≥95%
Sigma-Aldrich
α-Ketoglutaric acid disodium salt dihydrate, ≥98.0% (dried material, NT)