Skip to Content
Merck
All Photos(5)

Key Documents

K1875

Sigma-Aldrich

α-Ketoglutaric acid sodium salt

≥98% (titration)

Synonym(s):

2-Oxoglutaric acid monosodium salt, 2-Oxopentanedioic acid monosodium salt, Sodium 2-oxoglutarate monobasic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COCOONa
CAS Number:
Molecular Weight:
168.08
Beilstein:
4597521
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.28

Assay

≥98% (titration)

form

powder

solubility

water: 100 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[Na+].OC(=O)CCC(=O)C([O-])=O

InChI

1S/C5H6O5.Na/c6-3(5(9)10)1-2-4(7)8;/h1-2H2,(H,7,8)(H,9,10);/q;+1/p-1

InChI key

MOTOGHHLNTXPTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Li Chen et al.
Journal of molecular modeling, 19(4), 1919-1927 (2013-01-23)
Bacillus subtilis natto is widely used in industry to produce natto, a traditional and popular Japanese soybean food. However, during its secondary fermentation, high amounts of ammonia are released to give a negative influence on the flavor of natto. Glutamate
Ming-Ming Jin et al.
Molecular biology reports, 40(4), 2995-3002 (2012-12-25)
Microcystis aeruginosa is the key symptom of water eutrophication and produces persistent microcystins. Our special attention was paid to the isocitrate dehydrogenase (IDH) of M. aeruginosa (MaIDH) because it plays important roles in energy and biosynthesis metabolisms and its catalytic
E Tomaszewska et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 63(5), 547-554 (2012-12-06)
The potential effects of prenatal administration of dexamethasone (DEX) and postnatal treatment with 2-oxoglutaric acid (2-Ox) on postnatal development of connective tissue of farm animals were not examined experimentally. The aim of this study was to establish changes in morphological
Piotr Dobrowolski et al.
Nutrition (Burbank, Los Angeles County, Calif.), 29(3), 556-561 (2012-12-12)
Proton-pump inhibitors, such as omeprazole, are widely used in the prevention and treatment of gastroesophageal diseases. However, an association between proton-pump inhibitors and the increased risk of bone fractures has been observed, especially in patients treated for extended periods. Conversely
Michael E Jung et al.
The Journal of organic chemistry, 77(23), 11002-11005 (2012-11-21)
Oxidative cleavage of cycloalkene-1-carboxylates, made from the corresponding carboxylic acids, and subsequent oxidation of the resulting ketoaldehyde afforded the important 1-monoesters of 2-ketoalkanedioic acids. Thus ozonolysis of octyl cyclobutene-1-carboxylate followed by sodium chlorite oxidation afforded the 1-monooctyl 2-ketoglutarate. This is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service