Merck
  • Extracts from the Cochrane Library: interventions for the prevention of postoperative ear discharge after insertion of ventilation tubes (grommets) in children.
Extracts from the Cochrane Library: interventions for the prevention of postoperative ear discharge after insertion of ventilation tubes (grommets) in children.
Otolaryngology--head and neck surgery : official journal of American Academy of Otolaryngology-Head and Neck Surgery (2013-11-22)
Anne G M Schilder, Martin J Burton, Jennifer J Shin, Richard M Rosenfeld
ABSTRACT
The "Cochrane Corner" is a section in the journal that highlights systematic reviews relevant to otolaryngology-head and neck surgery, with invited commentary to aid clinical decision making. This installment features a Cochrane Review, "Interventions for the Prevention of Postoperative Ear Discharge after Insertion of Ventilation Tubes (Grommets) in Children," that concludes if a surgeon has a high rate of postoperative otorrhea in children, then saline irrigation or antibiotic ear drops at the time of surgery would significantly reduce that rate. If topical drops are chosen, a single application at the time of surgery is suggested to reduce the cost and potential for ototoxic damage (if applicable).
Sigma-Aldrich
Chloride standard solution, 1 mg/mL Cl-
Sigma-Aldrich
Sodium standard solution, suitable for atomic absorption spectrometry, 1000 ppm Na, 1 mg/mL Na
Sigma-Aldrich
Sodium chloride solution, 0.1 M
Sigma-Aldrich
Sodium chloride, JIS special grade, ≥99.5%
Sigma-Aldrich
Sodium chloride, SAJ first grade, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 1 M
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.