Merck
  • Production and evaluation of biodiesel and bioethanol from high oil corn using three processing routes.
Production and evaluation of biodiesel and bioethanol from high oil corn using three processing routes.
Bioresource technology (2012-12-01)
Aimin Shi, Zhenyi Du, Xiaochen Ma, Yanling Cheng, Min Min, Shaobo Deng, Paul Chen, Dong Li, Roger Ruan
ABSTRACT
Six Korea high oil (KHO) corn varieties varying in germ and endosperm size and oil content (4-21%, wet basis) were subjected to three sequential combinations of milling (M), germ separation (S), fermentation (F), and in situ transesterification (T) to produce bioethanol and biodiesel. Production parameters including saccharification, bioethanol yield, biodiesel yield and composition, and conversion rate were evaluated. The effects of the contents of germ, endosperm size, oil, and non-oil solid mass on the production parameters strongly depended on the processing routes, namely M-F-T, M-T-F, and S-T|F. The M-F-T route produced the highest bioethanol yield while the S-T|F route produced the highest biodiesel yield. The in situ transesterification reaction, if proceeded before fermentation, reduced the bioethanol yield while fermentation and/or presence of endosperm reduced the biodiesel yield.
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Supelco
Corn oil, analytical standard
Sigma-Aldrich
Corn oil, CP