Merck
  • Anterior and posterior partial-coverage indirect restorations using supragingival dentistry techniques.
Anterior and posterior partial-coverage indirect restorations using supragingival dentistry techniques.
Journal of the Massachusetts Dental Society (2013-03-22)
Jose-Luis Ruiz
ABSTRACT
The literature is replete with articles arguing which is the next best restorative material to substitute porcelain fused to metal (PFM) crowns. Sadly, most of the discussion is about the strength and cost of the materials and often not about how--when properly used--some of these materials, combined with the appropriate techniques, can actually improve the quality of the dentistry we provide to our patients and make our restorative life easier and more predictable.
Sigma-Aldrich
Poly(methyl methacrylate), CP
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic