Merck
  • A facile and versatile approach to design self-assembled monolayers on glass using thiol-ene chemistry.
A facile and versatile approach to design self-assembled monolayers on glass using thiol-ene chemistry.
Chemical communications (Cambridge, England) (2013-01-23)
Birgit Oberleitner, Antoine Dellinger, Maxime Déforet, Anouk Galtayries, Anne-Sophie Castanet, Vincent Semetey
ABSTRACT
This work describes an integrated approach for designing on demand Self-Assembled Monolayers (SAMs) on silicon oxides and particularly glass substrates for cell biology applications. Starting from commercially available compounds, the strategy relies on thiol-ene reaction and provides high quality SAMs exhibiting adhesive and anti-adhesive patterns.
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Ethylene glycol, SAJ first grade, ≥99.0%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylene glycol solution, NMR reference standard, 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Supelco
Ethylene glycol, analytical standard
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylene glycol, JIS special grade, ≥99.5%
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol 5 M solution