Merck
  • Lipid droplet synthesis is limited by acetate availability in starchless mutant of Chlamydomonas reinhardtii.
Lipid droplet synthesis is limited by acetate availability in starchless mutant of Chlamydomonas reinhardtii.
FEBS letters (2013-01-15)
Rishiram Ramanan, Byung-Hyuk Kim, Dae-Hyun Cho, So-Ra Ko, Hee-Mock Oh, Hee-Sik Kim
ABSTRACT
Phenotypic and genotypic changes in Chlamydomonas reinhardtii BafJ5, a starchless mutant, with respect to lipid metabolism was studied in different trophic states under nitrogen (N) sufficient and limited conditions. Interestingly, cellular lipid content increased linearly with input acetate concentration with highest lipid content (∼42%) under nitrogen limitation and mixotrophic state. RT-qPCR studies indicate that key fatty acid biosynthesis genes are down-regulated under N limitation but not under mixotrophic state, whereas, ACS2, encoding Acetyl-CoA synthetase, and DGTT4, encoding Diacylglycerol O-acyltransferase, are up-regulated under all conditions. These results collectively indicate that acetate is the limiting factor and central molecule in lipid droplet synthesis. The study also provides further evidence of the presence of a chloroplast pathway for triacylglycerol synthesis in microalgae.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%