Merck
  • C(sp3)-F bond activation of CF3-substituted anilines with catalytically generated silicon cations: spectroscopic evidence for a hydride-bridged Ru-S dimer in the catalytic cycle.
C(sp3)-F bond activation of CF3-substituted anilines with catalytically generated silicon cations: spectroscopic evidence for a hydride-bridged Ru-S dimer in the catalytic cycle.
Journal of the American Chemical Society (2013-01-15)
Timo Stahl, Hendrik F T Klare, Martin Oestreich
ABSTRACT
Heterolytic splitting of the Si-H bond mediated by a Ru-S bond forms a sulfur-stabilized silicon cation that is sufficiently electrophilic to abstract fluoride from CF(3) groups attached to selected anilines. The ability of the Ru-H complex, generated in the cooperative activation step, to intramolecularly transfer its hydride to the intermediate carbenium ion (stabilized in the form of a cationic thioether complex) is markedly dependent on the electronic nature of its phosphine ligand. An electron-deficient phosphine thwarts the reduction step but, based on the Ru-S catalyst, half of an equivalent of an added alkoxide not only facilitates but also accelerates the catalysis. The intriguing effect is rationalized by the formation of a hydride-bridged Ru-S dimer that was detected by (1)H NMR spectroscopy. A refined catalytic cycle is proposed.
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%