Merck
  • Non-natural elemane as the "stepping stone" for the synthesis of germacrane and guaiane sesquiterpenes.
Non-natural elemane as the "stepping stone" for the synthesis of germacrane and guaiane sesquiterpenes.
Organic letters (2012-12-22)
Elissavet E Anagnostaki, Alexandros L Zografos
ABSTRACT
The synthesis of hydroxyelemane 5 from (R)-carvone and its utilization as a common synthetic scaffold to produce structurally diverse germacrane and guaiane sesquiterpenes are described. A highly enantio- and stereoselective biomimetic tandem oxy-Cope/ene rearrangement was used as the key reaction to access the 10-membered macrocyclic core of germacranes and the condensed 5-7 carbocycles of guaiane sesquiterpenes. Additionally, reactions of furanoguaianes under acidic or oxidizing reagents have been investigated, and preliminary results of these conversions are presented.
Sigma-Aldrich
L-Carvone, ≥97%, FCC, FG
Sigma-Aldrich
L-Carvone, natural, 99%, FG
Supelco
(+)-Carvone, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
(S)-(+)-Carvone, 96%
Sigma-Aldrich
(R)-(−)-Carvone, 98%
Supelco
(+)-Carvone, analytical standard
Sigma-Aldrich
D-Carvone, ≥96%, FG
Supelco
(−)-Carvone, analytical standard