Skip to Content
Merck
All Photos(1)

Documents

W224908

Sigma-Aldrich

L-Carvone

natural, 99%, FG

Synonym(s):

(R)-(−)-Carvone, (−)-Carvone, (R)-5-Isopropenyl-2-methyl-2-cyclohexenone, p-Mentha-6,8-dien-2-one, Carvol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14O
CAS Number:
Molecular Weight:
150.22
FEMA Number:
2249
Beilstein:
2206714
EC Number:
Council of Europe no.:
146
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
7.012
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

vapor density

5.2 (vs air)

vapor pressure

0.4 mmHg ( 20 °C)

Assay

99%

composition

contains IFRA restricted carvone

refractive index

n20/D 1.497 (lit.)

bp

227-230 °C (lit.)

density

0.959 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

carvone

Organoleptic

herbaceous; spearmint

SMILES string

CC(=C)[C@@H]1CC=C(C)C(=O)C1

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9H,1,5-6H2,2-3H3/t9-/m1/s1

InChI key

ULDHMXUKGWMISQ-SECBINFHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Carvone is one of the key constituents of spearmint (Mentha spicata) oil. Studies indicate that it is a potent feeding-deterrent to stored product beetles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Plant compounds that induce polychlorinated biphenyl biodegradation by Arthrobacter sp. strain B1B.
Gilbert ES & Crowley DE.
Applied and Environmental Microbiology, 63(5), 1933-1938 (1997)
Bioactivities of l-carvone, d-carvone, and dihydrocarvone toward three stored product beetles
Tripathi AK, et al.
Journal of Economic Entomology, 96(5), 1594-1601 (2003)
Carvone: Why and how should one bother to produce this terpene.
de Carvalho CC & da Fonseca MMR.
Food Chemistry, 95(3), 413-422 (2006)
Tingting Li et al.
Frontiers in microbiology, 9, 3324-3324 (2019-01-29)
Quorum sensing (QS), one of the most remarkable microbiological discoveries, is considered a global gene regulatory mechanism for various traits in bacteria, including virulence and spoilage. Hafnia alvei, an opportunistic pathogen and a dominant psychrophile, uses the lux-type QS system
Raul Conde et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 18(14), 1197-1201 (2011-08-02)
There is no universally accepted and effective prophylaxis of migraine headache episodes. Thus we aimed to investigate the effects of Lippia alba (Mill.) N. E. Brown, an herb with many effects on central nervous system, on pain frequency and intensity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service