Merck
  • Iron-catalyzed chemo- and stereoselective hydromagnesiation of diarylalkynes and diynes.
Iron-catalyzed chemo- and stereoselective hydromagnesiation of diarylalkynes and diynes.
Journal of the American Chemical Society (2012-10-05)
Laurean Ilies, Takumi Yoshida, Eiichi Nakamura
ABSTRACT
Diarylalkynes are chemo- and stereoselectively hydromagnesiated in high yields at room temperature with an iron species generated in situ from FeCl(2)and EtMgBr. Functional groups such as bromide, iodide, amine, phenoxide, and alkene are well tolerated. Under similar conditions, diynes are chemo-, regio-, and stereoselectively hydromagnesiated. The resulting alkenylmagnesium compounds are a platform for further functionalization as a one-pot reaction.
Sigma-Aldrich
Iron(II) chloride tetrahydrate, SAJ first grade, 98.0-102.0%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%