Merck
  • Graphene-SnO2 composites for highly efficient photocatalytic degradation of methylene blue under sunlight.
Graphene-SnO2 composites for highly efficient photocatalytic degradation of methylene blue under sunlight.
Nanotechnology (2012-08-17)
Humaira Seema, K Christian Kemp, Vimlesh Chandra, Kwang S Kim
ABSTRACT
Graphene sheets decorated with SnO(2) nanoparticles (RGO-SnO(2)) were prepared via a redox reaction between graphene oxide (GO) and SnCl(2). Graphene oxide (GO) was reduced to graphene (RGO) and Sn(2+) was oxidized to SnO(2) during the redox reaction, leading to a homogeneous distribution of SnO(2) nanoparticles on RGO sheets. The scanning electron microscopy (SEM) and transmission electron microscopy (TEM) images show uniform distribution of the nanoparticles on the RGO surface and high-resolution transmission electron microscopy (HRTEM) shows an average particle size of 3-5 nm. The RGO-SnO(2) composite showed an enhanced photocatalytic degradation activity for the organic dye methylene blue under sunlight compared to bare SnO(2) nanoparticles. This result leads us to believe that the RGO-SnO(2) composite could be used in catalytic photodegradation of other organic dyes.
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.97% trace metals basis
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride, anhydrous, powder, ≥99.99% trace metals basis
Supelco
Tin(II) chloride dihydrate, suitable for AAS
Sigma-Aldrich
Tin(II) chloride, reagent grade, 98%
Sigma-Aldrich
Tin(II) chloride dihydrate, ACS reagent, 98%
Sigma-Aldrich
Tin(II) chloride dihydrate, reagent grade, 98%
Sigma-Aldrich
Tin(II) chloride dihydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Tin standard solution, suitable for atomic absorption spectrometry, 1000 ppm Sn, 1 mg/mL Sn