Skip to Content
Merck
All Photos(1)

Key Documents

452335

Sigma-Aldrich

Tin(II) chloride

anhydrous, powder, ≥99.99% trace metals basis

Synonym(s):

Stannous chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnCl2
CAS Number:
Molecular Weight:
189.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
form:
powder
solubility:
water: soluble 1780 g/L at 10 °C

grade

anhydrous

Quality Level

vapor pressure

33 hPa (~429 °C)

Assay

≥99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: tin

pH

2.18 (20 °C)

bp

606 °C
652 °C (lit.)

mp

246 °C (lit.)

solubility

water: soluble 1780 g/L at 10 °C

SMILES string

Cl[SnH2]Cl

InChI

1S/2ClH.Sn/h2*1H;/q;;+2/p-2

InChI key

AXZWODMDQAVCJE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tin(II) chloride (SnCl2), also known as stannous chloride, is commonly used as a reducing agent.

Application

Synthesis of tin(IV) octaethylcorroles was accomplished with this reagent. These new compounds exhibit reversible oxidation only at the conjugated ring system, not at the metal center.
Tin(II) chloride (SnCl2) has been used to label compounds with 99mtechnetium (99mTc).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Cardio-vascular system, Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karl M. Kadish et al.
Inorganic chemistry, 37(18), 4573-4577 (2001-10-24)
Two Sn(IV) corroles were synthesized and electrochemically examined. The investigated compounds are represented as (OEC)Sn(C(6)H(5)) and (OEC)SnCl, where OEC = trianion of 2,3,7,8,12,13,17,18-octaethylcorrole. (OEC)Sn(C(6)H(5)) represents the first example of a sigma-bonded metallocorrole which does not undergo a metal-centered electrode reaction.
Seung Il Lee et al.
Scientific reports, 9(1), 2411-2411 (2019-02-23)
Blue phosphorescent organic light-emitting diodes (PHOLEDs) were fabricated with tin oxide (SnOx) nano-particles (NPs) deposited at the ITO anode to improve their electrical and optical performances. SnOx NPs helped ITO to increase the work function enhancing hole injection capability. Charge
Jae-Ho Lee et al.
Journal of liposome research, 23(4), 336-342 (2013-07-25)
The radiolabeling of the liposome surface can be a useful tool for in vivo tracking of therapeutic drug loaded liposomes. We investigated radiolabeling therapeutic drug (i.e. an antibiotic, amikacin) loaded liposomes with (99m)Tc, nebulization properties of (99m)Tc-labeled liposomal amikacin for
Tobias Bubmann et al.
Polymers, 11(12) (2019-12-18)
The effect of different catalysts on reactive compatibilization of 50/50 polycarbonate (PC)/polymethylmethacrylate (PMMA) blends achieved via transesterification that occurs during compounding in a twin-screw extruder was investigated on a phenomenological (optical and mechanical properties), mesoscopic (phase morphology), and molecular level
Jae-Ho Lee et al.
Journal of liposome research, 25(2), 101-106 (2014-06-26)
Porphyrin-lipid nanovesicles (PLN) have been developed with intrinsic capabilities as activatable multimodal photonic contrast agents. Radiolabeling of PLN encapsulating drugs could eventually be able to provide quantitative in vivo information for diagnosing and treating diseases. In this study, we developed

Articles

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Silylethyne substitution offers an opportunity to tune solubility for application-specific needs and self-assembly for electronic performance and has yielded semiconductors with excellent device performance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service