Merck
  • Evaluation of effectiveness of EDTA and sodium thiosulfate in removing metal toxicity toward sea urchin embryo-larval applying the TIE.
Evaluation of effectiveness of EDTA and sodium thiosulfate in removing metal toxicity toward sea urchin embryo-larval applying the TIE.
Chemosphere (2012-05-26)
C Resgalla, F Poleza, R C Souza, M V Máximo, C M Radetski
ABSTRACT
Since the development of the TIE (Toxicity Identification and Evaluation) in 1988 it has been assumed that the capacity of EDTA and sodium thiosulfate to complex some metals, and thus remove their toxicity, can be applied to both freshwater and seawater ecotoxicological tests and the results subsequently interpreted. However, it is now known that there is a wide variability in the extent of this complexation. In this context, the removal of toxicity caused by the presence of Hg(2+), Cd(2+), Cu(2+), Cr(6+), Zn(2+), Ni(2+), Pb(2+), Ag(1+) and Se(2+), through metal complexation by EDTA and sodium thiosulfate, in relation to the performance of embryo-larval tests with the sea urchin Arbacia lixula was investigated. It was observed that EDTA was capable of removing the toxicity of Pb(2+), Zn(2+) and Cu(2+) while sodium thiosulfate only reduced the toxicity of Ag(1+). Compared to the complexation observed in freshwater ecotoxicological tests, the complexing agents used in this study (EDTA and sodium thiosulfate) have a lower capacity to complex metals in the marine ecotoxicological test with A. lixula.
Sigma-Aldrich
Sodium thiosulfate pentahydrate, 99.999% trace metals basis
Sigma-Aldrich
Sodium thiosulfate pentahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate solution, 1 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.025 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.01 M
Sigma-Aldrich
Sodium thiosulfate solution, 0.1 M
Sigma-Aldrich
Sodium thiosulfate pentahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate pentahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium thiosulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium thiosulfate, purum p.a., anhydrous, ≥98.0% (RT)
Sigma-Aldrich
Sodium thiosulfate, ReagentPlus®, 99%