Skip to Content
Merck
All Photos(2)

Key Documents

S8503

Sigma-Aldrich

Sodium thiosulfate pentahydrate

ReagentPlus®, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

Agency

suitable for SM 2310
suitable for SM 2320

product line

ReagentPlus®

Assay

≥99.5%

form

powder

storage temp.

room temp

SMILES string

O.O.O.O.O.[Na+].[Na+].[O-]S([O-])(=O)=S

InChI

1S/2Na.H2O3S2.5H2O/c;;1-5(2,3)4;;;;;/h;;(H2,1,2,3,4);5*1H2/q2*+1;;;;;;/p-2

InChI key

PODWXQQNRWNDGD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Screening of Variables Affecting the Selective Leaching of Valuable Metals from Waste Motherboards′ PCBs: Demonstrates the use of sodium thiosulfate pentahydrate in the process of leaching valuable metals from electronic waste, contributing significantly to sustainable recycling practices in environmental science (Rahimi et al., 2024).

  • Synthesis of Aryl Amides from Acyl-Bunte Salts and Aryl Azides: Details the application of sodium thiosulfate pentahydrate in synthetic organic chemistry, specifically in the synthesis of aryl amides, illustrating its utility in facilitating complex chemical reactions (Chen et al., 2023).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christian Pfeifer et al.
International orthopaedics, 36(7), 1493-1499 (2012-01-13)
The purpose of this study was to find a suitable method of labelling cartilage samples for the measurement of distraction distances in biomechanical testing. Samples of bovine cartilage were labelled using five different methods: hydroquinone and silver nitrate (AgNO3), potassium
Melani Sooriyaarachchi et al.
Metallomics : integrated biometal science, 5(3), 197-207 (2013-02-28)
Cis-Platin (CP) is a remarkably effective Pt-based anticancer drug, but it also exhibits severe toxic side-effects, including nephrotoxicity and ototoxicity. Previous studies conducted with mammalian model organisms have clearly demonstrated that the intravenous administration of N-acetyl-L-cysteine (NAC) or sodium thiosulfate
C Resgalla et al.
Chemosphere, 89(1), 102-107 (2012-05-26)
Since the development of the TIE (Toxicity Identification and Evaluation) in 1988 it has been assumed that the capacity of EDTA and sodium thiosulfate to complex some metals, and thus remove their toxicity, can be applied to both freshwater and
Michael C Reade et al.
Emergency medicine Australasia : EMA, 24(3), 225-238 (2012-06-08)
Cyanide poisoning is uncommon, but generates interest because of the presumed utility of an antidote immediately available in those areas with a high risk of cyanide exposure. As part of its regular review of guidelines, the Australian Resuscitation Council conducted
Brooke Bair et al.
Journal of drugs in dermatology : JDD, 10(9), 1042-1044 (2011-11-05)
Calcinosis cutis is a term used to describe a group of disorders which result in calcium deposits in the skin. These disorders can be separated based on etiology. Sodium thiosulfate has been used to systemically treat calciphylaxis with little to

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service