Merck
  • Gap site-specific rapid formation of fluorescent silver nanoclusters for label-free DNA nucleobase recognition.
Gap site-specific rapid formation of fluorescent silver nanoclusters for label-free DNA nucleobase recognition.
Analytica chimica acta (2012-04-10)
Qinghua Cui, Kun Ma, Yong Shao, Shujuan Xu, Fei Wu, Guiying Liu, Norio Teramae, Haifeng Bao
ABSTRACT
Silver nanoclusters (Ag NCs) templated with DNAs have attracted much attention as novel fluorophores because of their convenient emission tunability by the sequence and length of the template DNAs. However, the precise production of Ag NCs in a site-specific manner still remains a challenge to attain highly selective and label-free DNA recognition. Here we exploited the availability of a gap site in DNA duplexes as a new scaffold for the synthesis of Ag NCs. Compared to the commonly used DNA templates for the creation of Ag NCs, the gap site in DNA duplexes was found to facilitate the rapid formation of the fluorescent Ag NCs without sacrifice of their bright emission and excellent stability. We found that fluorescent Ag NCs were highly selectively formed when cytosine faced toward the gap site in DNA duplexes, and they were in situ utilized as readout by signal-on manner for the DNA mutation assays. This base-selective growth of the fluorescent Ag NCs at the gap site would find promising applications in practical detection of single nucleotide polymorphism (SNP) and construction of DNA-based functional sensors with label-free and cost-effective merits.
Sigma-Aldrich
VenPure® SF, powder
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%