Skip to Content
Merck
All Photos(4)

Key Documents

452874

Sigma-Aldrich

Sodium borohydride

greener alternative

granular, 10-40 mesh, 98%

Synonym(s):

VenPure® AF, Sodium tetrahydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaBH4
CAS Number:
Molecular Weight:
37.83
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

granular

reaction suitability

reagent type: reductant

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

10-40 mesh

mp

>300 °C (dec.) (lit.)

greener alternative category

SMILES string

[BH4-].[Na+]

InChI

1S/BH4.Na/h1H4;/q-1;+1

InChI key

YOQDYZUWIQVZSF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Sodium borohydride (NaBH4) is a stable hydrogen carrier, which can be used as a chemical hydrogen storage material. It can also be used as an anodic fuel for alternative energy applications such as the fabrication of fuel cells. NaBH4 can also be used in the reduction of graphene oxide in the presence of CaCl2 as a catalyst.
Sodium borohydride can be used to prepare:
  • Silica-coated silver nanoparticles, which are functionalized with various vitamins and antibiotics as bioactive materials.
  • Water-soluble oligoamine conjugated chitosans for biomedical applications.
  • Gold nanoparticles coated with poly(methacrylic acid), which are used for theranostic applications.

Legal Information

Ascensus Specialties LLC
VenPure is a registered trademark of Ascensus Specialties LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 1B - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Preparation and characterization of iron-polypyrrole-carbon composite for use as methanol tolerant cathode material in direct methanol fuel cells.
Madhu CS and Singh RN
Indian J. Chem. A, 52(11), 1383-1390 (2013)
Ternary platinum-multiwall carbon nanotube-cobaltite composites for methanol electrooxidation.
Awasthi R and Singh RN
International Journal of Electrochemistry, 6, 4775-4775 (2011)
Poly (methacrylic acid)-coated gold nanoparticles: functional platforms for theranostic applications
Yilmaz G, et al.
Biomacromolecules, 17(9), 2901-2911 (2016)
Oligoamines conjugated chitosan derivatives: Synthesis, characterization, in vitro and in vivo biocompatibility evaluations
Jia Y, et al.
Carbohydrate Polymers, 83(3), 1153-1161 (2011)
Hydrogen generation using sodium borohydride solution and metal catalyst coated on metal oxide
Kojima Y, et al.
International Journal of Hydrogen Energy, 27(10), 1029-1034 (2002)

Articles

An article about metal borohydrides as hydrogen storage materials

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service