Merck
  • Phenolics composition and antioxidant activity of wine produced from spine grape (Vitis davidii Foex) and Cherokee rose (Rosa laevigata Michx.) fruits from South China.
Phenolics composition and antioxidant activity of wine produced from spine grape (Vitis davidii Foex) and Cherokee rose (Rosa laevigata Michx.) fruits from South China.
Journal of food science (2011-12-21)
Jiangfei Meng, Yulin Fang, Jinshan Gao, Lingling Qiao, Ang Zhang, Zhijun Guo, Minyang Qin, Jizhuang Huang, Ying Hu, Xifu Zhuang
ABSTRACT
Wines made from 3 spine grape (Vitis davidii Foex) genotypes-Junzi 1# (JZ 1#), Junzi 2# (JZ 2#), and Liantang (LT)-and Cherokee rose (Rosa laevigata Michx., CR) were evaluated for their phenolics composition and antioxidant activities by several assays, including 2,2'-azino-bis-(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt (ABTS scavenging capacity, cupric ion reducing antioxidant capacity, and metal-chelating capacity). The results showed that CR wine had higher contents of total phenolics, total flavonoids, and oligomeric proanthocyanidins than the 3 spine grape genotype wines and Cabernet Sauvignon (CS) wine. Among the 3 varieties of spine grape wines, JZ 1# had higher contents than the 2 other genotypes. Moreover, the total monomeric anthocyanins in JZ 1# was about 4.5-fold higher than those in CS wine and CR wine. A significance analysis demonstrated that the antioxidant capacity (as measured by the ABTS scavenging capacity and cupric ion reducing antioxidant capacity) of CR wine was significantly higher than that of the control and spine grape wines. With respect to metal-chelating capacity, the CR wine was the strongest amongst all the wine samples tested, followed by JZ 1#. This indicates that CR and JZ 1# wines can potentially be considered as wild fruit wines with abundant phenolic compounds and antioxidant activities. Spine grape (Vitis davidii Foex) and Cherokee rose (Rosa laevigata Michx.) are 2 species of important wild plants in South China. However, they have not yet been effectively developed and utilized. Recently, some researchers attempted to convert their juice to wine. We anticipate that this research will provide sufficient experimental evidence for their good antioxidant activity and potential for further development and utilization.
Sigma-Aldrich
Iron(II) chloride tetrahydrate, SAJ first grade, 98.0-102.0%
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.998% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Iron(II) chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride tetrahydrate, 99.99% trace metals basis
Sigma-Aldrich
Iron(II) chloride, 98%
Sigma-Aldrich
Iron(II) chloride tetrahydrate, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Iron(II) chloride tetrahydrate, ReagentPlus®, 98%