Merck
  • A computer Time Trade-Off: a feasible and reliable alternative for the interview Time Trade-Off in rheumatoid arthritis.
A computer Time Trade-Off: a feasible and reliable alternative for the interview Time Trade-Off in rheumatoid arthritis.
Clinical and experimental rheumatology (2011-10-04)
Laurien Buitinga, Louise M A Braakman-Jansen, Erik Taal, Mart A F J van de Laar
ABSTRACT
The Time Trade-Off (TTO) is an instrument used for valuing health-related quality of life. This study evaluated the test-retest reliability of a computer TTO in rheumatoid arthritis (RA), and compared the computer with the interview TTO regarding feasibility and agreement. In study 1 using a cross-over design, thirty patients completed both TTOs. In study 2, twenty-nine other patients completed the computer TTO twice to examine test-retest reliability. Feasibility was measured by assessing actual and perceived time duration and general experience of the patient. Agreement between utility scores of both TTOs was measured by Bland-Altman analysis. Both TTOs were feasible. The computer TTO showed high test-retest reliability (ICC = 0.88). Bland-Altman analysis showed a small mean difference (0.06, SD = 0.14, effect size=0.30) between both TTOs. Limits of agreement were wide (-0.22 to 0.34). Differences between interview and computer TTO utilities did not vary over the range of scores. The computer TTO was feasible and reliable, but did not provide similar results as the interview TTO. However, no systematic biases in the differences were found over the range of scores.
Sigma-Aldrich
Poloxamer 188 solution, 10%, sterile-filtered, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,000
Sigma-Aldrich
Poloxamer 188, micronized
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
Synperonic® F 108, surfactant, non-ionic
Sigma-Aldrich
Poloxamer 407, oxyethylene 71.5-74.9 %
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~8,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,100
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~4,400
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~1,900
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~2,000
Sigma-Aldrich
Synperonic® PE/P84
Sigma-Aldrich
Poloxamer 407, micronized
Sigma-Aldrich
Pluronic® F-127, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~14,600
Sigma-Aldrich
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol), average Mn ~3,300
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~2,900
Sigma-Aldrich
Synperonic® PE P105, surfactant
Sigma-Aldrich
Poloxamer 188, solid
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), average Mn ~5,800
Sigma-Aldrich
Poloxamer 188