Merck
  • Tantalum compounds as heterogeneous catalysts for saccharide dehydration to 5-hydroxymethylfurfural.
Tantalum compounds as heterogeneous catalysts for saccharide dehydration to 5-hydroxymethylfurfural.
Chemical communications (Cambridge, England) (2011-03-04)
Fengli Yang, Qishun Liu, Min Yue, Xuefang Bai, Yuguang Du
ABSTRACT
A new solid acid, based on tantalum hydroxide, was used to catalyze saccharide dehydration into 5-hydroxymethylfurfural (HMF) with high catalytic activity and excellent stability in a water-2-butanol biphasic system. Furthermore, good results were also obtained from Jerusalem artichoke juice with the catalyst under the same conditions.
Sigma-Aldrich
2-Butanol, JIS special grade, ≥99.0%
Sigma-Aldrich
2-Butanol, anhydrous, 99.5%
Sigma-Aldrich
Aluminum-tri-sec-butoxide, 97%
Sigma-Aldrich
2-Butanol, ReagentPlus®, ≥99%
Supelco
2-Butanol, analytical standard
Sigma-Aldrich
(S)-(+)-2-Butanol, 99%