Skip to Content
Merck
All Photos(1)

Documents

237698

Sigma-Aldrich

(S)-(+)-2-Butanol

99%

Synonym(s):

(S)-(+)-sec-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5CH(OH)CH3
CAS Number:
Molecular Weight:
74.12
Beilstein:
1718763
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

vapor density

2.6 (vs air)

Quality Level

vapor pressure

12.5 mmHg ( 20 °C)

Assay

99%

form

liquid

optical activity

[α]20/D +13°, neat

autoignition temp.

761 °F

expl. lim.

9.8 %

refractive index

n20/D 1.397 (lit.)

bp

99-100 °C (lit.)

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC[C@H](C)O

InChI

1S/C4H10O/c1-3-4(2)5/h4-5H,3H2,1-2H3/t4-/m0/s1

InChI key

BTANRVKWQNVYAZ-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(+)-2-Butanol can be used:
  • To synthesize chiral asymmetric perylene diimides (PDI) that spontaneously self-assemble into one-handed nanotubes that are photoconductive or fluorescent in nature.
  • To synthesize bulky chiral vinyl monomers that can be radically polymerized to helical polymers with an excess screw sense.
  • As chiral solvent to study excited state photo transfer of the photoacid, 5,8-dicyano-2-naphthol (DCN2) in chiral environment.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Excited-state proton transfer in chiral environments. 1. Chiral solvents.
Solntsev KM, et al.
Journal of the American Chemical Society, 124(31), 9046-9047 (2002)
Highly fluorescent one-handed nanotubes assembled from a chiral asymmetric perylene diimide.
Ma X , et al.
Chemical Communications (Cambridge, England), 51(20), 4231-4233 (2015)
Stepwise Formation of Photoconductive Nanotubes through a New Top?Down Method.
Ma X , et al.
Advanced Materials, 27(47), 7746-7751 (2015)
Synthesis and characterization of optically active helical vinyl polymers via free radical polymerization.
Zhi J, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 47(9), 2408-2421 (2009)
Dagmara Wojtków et al.
The journal of physical chemistry. A, 110(36), 10552-10557 (2006-09-08)
The effect of temperature and concentration on the structure of sec-butyl alcohol and isobutyl alcohol/water binary mixtures in the alcohol-rich region (mole fraction of water X(H2O) < 0.3) has been studied using Fourier transform (FT) near-infrared (NIR) spectroscopy. The experimental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service