Merck
  • Design and optimization of fermentation medium for enhanced bacteriocin production by probiotic bacterium Enterococcus faecium MC13.
Design and optimization of fermentation medium for enhanced bacteriocin production by probiotic bacterium Enterococcus faecium MC13.
Preparative biochemistry & biotechnology (2011-01-14)
Paulraj Kanmani, R Satish Kumar, N Yuvaraj, K A Paari, V Pattukumar, Venkatesan Arul
ABSTRACT
Statistics-based experimental designs were used to develop a cost-effective medium for enhanced production of viable cells and bacteriocin by probiotic Enterococcus faecium MC13. Carbon, nitrogen, and mineral sources were first screened by one-variable-at-a-time (OVAT) methods. In order to increase yield production, the selected variables were further statistically optimized using response-surface methodology (RSM) with central composite design (CCD). The maximum and minimum levels of the selected variables were determined and a set of 34 experimental runs was performed. The optimum concentrations of the tested variables for production of viable cells (12.24 log CFU mL(-1)) and bacteriocin activity (25,600 AU mL(-1)) were tryptone (10.0 g/L), peptone (6.0 g/L), maltose (3.0 g/L), glucose (9.0 g/L), NaCl (15.0 g/L), sodium citrate (2.5 g/L), sodium acetate (1.0 g/L), and dipotassium PO(4) (0.1 g/L). Threefold increased yield of bacteriocin was achieved in optimized medium compared to the unoptimized counterpart, and this was two times less cost than commercial MRS medium.
Sigma-Aldrich
Peptone from animal tissue, SAJ first grade
Millipore
Peptone from animal tissue, from meat
Millipore
Peptone from casein and other animal proteins, suitable for microbiology
Sigma-Aldrich
Peptone from animal tissue, from meat, BioReagent, suitable for cell culture, suitable for plant cell culture
Millipore
Peptone Primatone® RL, suitable for microbiology
Millipore
Proteose Peptone, Enzymatic hydrolysate
Millipore
Peptone from milk solids, Refined hydrolysate