Merck
  • Electrical and photoresponse properties of an intramolecular p-n homojunction in single phosphorus-doped ZnO nanowires.
Electrical and photoresponse properties of an intramolecular p-n homojunction in single phosphorus-doped ZnO nanowires.
Nano letters (2009-07-09)
Ping-Jian Li, Zhi-Min Liao, Xin-Zheng Zhang, Xue-Jin Zhang, Hui-Chao Zhu, Jing-Yun Gao, K Laurent, Y Leprince-Wang, N Wang, Da-Peng Yu
ABSTRACT
The single-crystal n-type and p-type ZnO nanowires (NWs) were synthesized via a chemical vapor deposition method, where phosphorus pentoxide was used as the dopant source. The electrical and photoluminescence studies reveal that phosphorus-doped ZnO NWs (ZnO:P NWs) can be changed from n-type to p-type with increasing P concentration. Furthermore, we report for the first time the formation of an intramolecular p-n homojunction in a single ZnO:P NW. The p-n junction diode has a high on/off current ratio of 2.5 x 10(3) and a low forward turn-on voltage of approximately 1.37 V. Finally, the photoresponse properties of the diode were investigated under UV (325 nm) excitation in air at room temperature. The high photocurrent/dark current ratio (3.2 x 10(4)) reveals that the diode has a potential as extreme sensitive UV photodetectors.
Sigma-Aldrich
Phosphorus pentoxide, desiccant, with moisture indicator
Sigma-Aldrich
Phosphorus pentoxide, powder, ACS reagent, ≥98.0%
Sigma-Aldrich
Phosphorus pentoxide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorus pentoxide, ReagentPlus®, 99%
Supelco
Phosphorus pentoxide, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus pentoxide, puriss. p.a., ACS reagent, ≥98.0% (T)