Merck
  • Gas chromatography-optical fiber detector for the speciation of aromatic hydrocarbons in confined areas.
Gas chromatography-optical fiber detector for the speciation of aromatic hydrocarbons in confined areas.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2008-08-12)
Lurdes I B Silva, Teresa A P Rocha-Santos, A C Duarte
ABSTRACT
An analytical method, based on separation with gas chromatography (GC) and detection with optical fiber (OF), was used for the separation, detection and quantification of benzene, toluene, ethylbenzene, p-xylene, m-xylene and o-xylene. The use of OF as a detector is based on the variations of the reflected optical power detected when the aromatic compounds eluted from the GC column are sorbed in a thin polymeric film on a single-mode OF. General figures of merit, such as the analytical time, analytical error and analytical performance of GC-OF were similar to those of the classical analytical methods, such as a gas chromatography-flame ionization detector (GC-FID). However, the developed GC-OF method constitutes a much less expensive alternative for the speciation of aromatic hydrocarbons compounds, with high accuracy, and being most suitable for actual monitoring work on confined environments.
Product Description
Supelco
ORBO 32 Small Activated Coconut Charcoal (20/40), 100/50 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
m-Xylene, analytical standard
Sigma-Aldrich
m-Xylene, anhydrous, ≥99%
Supelco
m-Xylene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
m-Xylene, ReagentPlus®, 99%
Sigma-Aldrich
m-Xylene, puriss. p.a., ≥99.0% (GC)