Skip to Content
Merck
All Photos(1)

Documents

95670

Supelco

m-Xylene

analytical standard

Synonym(s):

1,3-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
605441
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.7 (vs air)

Assay

≥99.5% (GC)

autoignition temp.

982 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

7 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.497 (lit.)
n20/D 1.497

bp

138-139 °C (lit.)

mp

−48 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Cc1cccc(C)c1

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3

InChI key

IVSZLXZYQVIEFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Xylene is an aromatic liquid , which is an isomeric form of xylene.
m-Xylene is an aromatic liquid, which is an isomeric form of xylene.
m-Xylene is an environmental pollutant belonging to the class of volatile organic compounds. It is emitted from a variety of sources including adhesives, degreasing agents, combustion products of wood and fuels, industrial paints, aerosols, etc.

Application

m-Xylene may be used as an analytical reference standard for the quantification of the analyte in occupational environmental samples and groundwater samples using gas chromatography technique with different modes of detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Toxics A to Z: A Guide to Everyday Pollution Hazards (1991)
Toxics A to Z: A Guide to Everyday Pollution Hazards (1991)
Sittig's Handbook of Toxic and Hazardous Chemicals and Carcinogens (2017)
Sittig's Handbook of Toxic and Hazardous Chemicals and Carcinogens (2017)
Fingerprint identification of volatile organic compounds in gasoline contaminated groundwater using gas chromatography differential ion mobility spectrometry
Liang F, et al.
International Journal for Ion Mobility Spectrometry, 15(3), 169-177 (2012)

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service