Merck
  • Control of transdermal permeation of hydrocortisone acetate from hydrophilic and lipophilic formulations.
Control of transdermal permeation of hydrocortisone acetate from hydrophilic and lipophilic formulations.
AAPS PharmSciTech (2008-06-20)
Adamo Fini, Valentina Bergamante, Gian Carlo Ceschel, Celestino Ronchi, Carlos Alberto Fonseca De Moraes
ABSTRACT
The purpose of this research was the preparation of four formulations containing hydrocortisone acetate (HCA) for topical application, including two aqueous systems (hydrophilic microemulsion and aqueous gel) and two systems with dominant hydrophobicity (hydrophobic microemulsion and ointment). The formulations were tested for the release and permeation of HCA across an animal membrane. The release of HCA was found comparable for the four systems. The two microemulsions promote permeation across an ex-vivo membrane, examined by means of a Franz cell. Hydrophobic microemulsion guarantees the highest solubility (2,370 microg/ml) and flux (133 microg/cm(2).h) of the drug, since it contains almost 40% Transcutol, a permeation enhancer. Gel and ointment provide lower solubility and flux, being the values, related to the ointment, the lowest ones (562 microg/ml and 0.4 microg/cm(2).h). Experimental results allow the conclusion that gel and ointment can be suitable when it is desirable to minimize absorption of topically applied HCA as to keep the drug restricted to the diseased area and prevent side effects of the systemic presence of HCA.
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Hydrocortisone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrocortisone 21-acetate, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone 21-acetate, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Supelco
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cortisol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
USP
Hydrocortisone acetate, United States Pharmacopeia (USP) Reference Standard
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
Hydrocortisone acetate, European Pharmacopoeia (EP) Reference Standard