Merck
  • Astrocyte growth is driven by the Tre1/S1pr1 phospholipid-binding G protein-coupled receptor.
Astrocyte growth is driven by the Tre1/S1pr1 phospholipid-binding G protein-coupled receptor.
Neuron (2023-12-15)
Jiakun Chen, Tobias Stork, Yunsik Kang, Katherine A M Nardone, Franziska Auer, Ryan J Farrell, Taylor R Jay, Dongeun Heo, Amy Sheehan, Cameron Paton, Katherine I Nagel, David Schoppik, Kelly R Monk, Marc R Freeman
ABSTRACT
Astrocytes play crucial roles in regulating neural circuit function by forming a dense network of synapse-associated membrane specializations, but signaling pathways regulating astrocyte morphogenesis remain poorly defined. Here, we show the Drosophila lipid-binding G protein-coupled receptor (GPCR) Tre1 is required for astrocytes to establish their intricate morphology in vivo. The lipid phosphate phosphatases Wunen/Wunen2 also regulate astrocyte morphology and, via Tre1, mediate astrocyte-astrocyte competition for growth-promoting lipids. Loss of s1pr1, the functional analog of Tre1 in zebrafish, disrupts astrocyte process elaboration, and live imaging and pharmacology demonstrate that S1pr1 balances proper astrocyte process extension/retraction dynamics during growth. Loss of Tre1 in flies or S1pr1 in zebrafish results in defects in simple assays of motor behavior. Tre1 and S1pr1 are thus potent evolutionarily conserved regulators of the elaboration of astrocyte morphological complexity and, ultimately, astrocyte control of behavior.
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Glutamine Synthetase (GS-6) Mouse Monoclonal Antibody
Roche
Anti-Digoxigenin-AP, Fab fragments, from sheep