Merck
  • Influence of dTMP on the phenotypic appearance and intracellular persistence of Staphylococcus aureus.
Influence of dTMP on the phenotypic appearance and intracellular persistence of Staphylococcus aureus.
Infection and immunity (2007-12-28)
Johannes Zander, Silke Besier, Stephan H Saum, Faramarz Dehghani, Stefan Loitsch, Volker Brade, Thomas A Wichelhaus
ABSTRACT
Thymidine-dependent small-colony variants (SCVs) of Staphylococcus aureus are frequently associated with persistent and recurrent infections in cystic fibrosis patients. The phenotypic appearance of S. aureus SCVs or normal-colony variants (NCVs) is postulated to be affected by the intracellular amount of dTMP. This hypothesis was proven by metabolic pathway assays revealing altered intracellular dTMP concentrations, followed by investigation of the associated phenotype. Inhibition of the staphylococcal thymidylate synthase, which generated intracellular dTMP from dUMP, using 5-fluorouracil and co-trimoxazole resulted in an SCV phenotype. Inhibition of a nucleoside transporter, which provided the bacterial cell with extracellular thymidine, caused growth inhibition of SCVs. In turn, reversion of SCVs to NCVs was achieved by supplying extracellular dTMP. High-performance liquid chromatography additionally confirmed the intracellular lack of dTMP in SCVs, in contrast to NCVs. Moreover, the dTMP concentration is postulated to influence the intracellular persistence of S. aureus. Cell culture experiments with cystic fibrosis cells revealed that clinical and co-trimoxazole-induced SCVs with a diminished amount of dTMP showed significantly better intracellular persistence than NCVs. In conclusion, these results show that the dTMP concentration plays a key role in both the phenotypic appearance and the intracellular persistence of S. aureus.
Product Description
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Uridine, ≥99%
Sigma-Aldrich
Uridine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Uridine, BioUltra, ≥99%
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 300
Sigma-Aldrich
Acetonitrile, ≥99.8%, for residue analysis, JIS 1000
Sigma-Aldrich
Acetonitrile, ≥99.8%, suitable for HPLC
Sigma-Aldrich
Acetonitrile, SAJ first grade, ≥99.0%
Sigma-Aldrich
Acetonitrile, JIS special grade, ≥99.5%
Sigma-Aldrich
Acetonitrile, for chromatography
Sigma-Aldrich
Acetonitrile, for residue analysis, JIS 5000
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Chloramphenicol, Ready Made Solution, 100 mg/mL in ethanol: isopropanol (95:5), ≥98% (HPLC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%