Merck
  • Modular antibodies reveal DNA damage-induced mono-ADP-ribosylation as a second wave of PARP1 signaling.
Modular antibodies reveal DNA damage-induced mono-ADP-ribosylation as a second wave of PARP1 signaling.
Molecular cell (2023-04-29)
Edoardo José Longarini, Helen Dauben, Carolina Locatelli, Anne R Wondisford, Rebecca Smith, Charlotte Muench, Andreas Kolvenbach, Michelle Lee Lynskey, Alexis Pope, Juan José Bonfiglio, Eva Pinto Jurado, Roberta Fajka-Boja, Thomas Colby, Marion Schuller, Ivan Ahel, Gyula Timinszky, Roderick J O'Sullivan, Sébastien Huet, Ivan Matic
ABSTRACT
PARP1, an established anti-cancer target that regulates many cellular pathways, including DNA repair signaling, has been intensely studied for decades as a poly(ADP-ribosyl)transferase. Although recent studies have revealed the prevalence of mono-ADP-ribosylation upon DNA damage, it was unknown whether this signal plays an active role in the cell or is just a byproduct of poly-ADP-ribosylation. By engineering SpyTag-based modular antibodies for sensitive and flexible detection of mono-ADP-ribosylation, including fluorescence-based sensors for live-cell imaging, we demonstrate that serine mono-ADP-ribosylation constitutes a second wave of PARP1 signaling shaped by the cellular HPF1/PARP1 ratio. Multilevel chromatin proteomics reveals histone mono-ADP-ribosylation readers, including RNF114, a ubiquitin ligase recruited to DNA lesions through a zinc-finger domain, modulating the DNA damage response and telomere maintenance. Our work provides a technological framework for illuminating ADP-ribosylation in a wide range of applications and biological contexts and establishes mono-ADP-ribosylation by HPF1/PARP1 as an important information carrier for cell signaling.
Sigma-Aldrich
Anti-SEPT2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-pan-ADP-ribose binding reagent, from Escherichia coli
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-poly-ADP-ribose binding reagent, Anti-poly-ADP-ribose binding reagent is a reagent that selectively binds to ADP ribose for use in Western Blotting, Immunocytochemistry and Dot Blot.
Sigma-Aldrich
Anti-unmodified-Histone H3 (Lys9) antibody, Mouse monoclonal, clone 9B1-2G6, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-RNF114 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Rabbit IgG HRP Linked Whole Ab, Cytiva NA934-1ML
Sigma-Aldrich
Anti-HPF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution