Merck
  • Optogenetic actuator - ERK biosensor circuits identify MAPK network nodes that shape ERK dynamics.
Optogenetic actuator - ERK biosensor circuits identify MAPK network nodes that shape ERK dynamics.
Molecular systems biology (2022-06-14)
Coralie Dessauges, Jan Mikelson, Maciej Dobrzyński, Marc-Antoine Jacques, Agne Frismantiene, Paolo Armando Gagliardi, Mustafa Khammash, Olivier Pertz
ABSTRACT
Combining single-cell measurements of ERK activity dynamics with perturbations provides insights into the MAPK network topology. We built circuits consisting of an optogenetic actuator to activate MAPK signaling and an ERK biosensor to measure single-cell ERK dynamics. This allowed us to conduct RNAi screens to investigate the role of 50 MAPK proteins in ERK dynamics. We found that the MAPK network is robust against most node perturbations. We observed that the ERK-RAF and the ERK-RSK2-SOS negative feedback operate simultaneously to regulate ERK dynamics. Bypassing the RSK2-mediated feedback, either by direct optogenetic activation of RAS, or by RSK2 perturbation, sensitized ERK dynamics to further perturbations. Similarly, targeting this feedback in a human ErbB2-dependent oncogenic signaling model increased the efficiency of a MEK inhibitor. The RSK2-mediated feedback is thus important for the ability of the MAPK network to produce consistent ERK outputs, and its perturbation can enhance the efficiency of MAPK inhibitors.
Sigma-Aldrich
SU-5402, ≥98% (HPLC)
Sigma-Aldrich
hBFGF, FGF-Basic, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, 351-368) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
RSK Inhibitor, SL0101, The RSK Inhibitor, SL0101 controls the biological activity of RSK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.