Merck
  • Carfilzomib modulates tumor microenvironment to potentiate immune checkpoint therapy for cancer.
Carfilzomib modulates tumor microenvironment to potentiate immune checkpoint therapy for cancer.
EMBO molecular medicine (2021-12-14)
Qian Zhou, Jinxia Liang, Tong Yang, Jin Liu, Bo Li, Yingchang Li, Zhenzhen Fan, Weida Wang, Wensheng Chen, Sujing Yuan, Meng Xu, Qigui Xu, Zhidong Luan, Zhongjun Xia, Penghui Zhou, Yadong Huang, Liang Chen
ABSTRACT
Impressive clinical benefit is seen in clinic with PD-1 inhibitors on portion of cancer patients. Yet, there remains an urgent need to develop effective synergizers to expand their clinical application. Tumor-associated macrophage (TAM), a type of M2-polarized macrophage, eliminates or suppresses T-cell-mediated anti-tumor responses. Transforming TAMs into M1 macrophages is an attractive strategy of anti-tumor therapy. Here, we conducted a high-throughput screening and found that Carfilzomib potently drove M2 macrophages to express M1 cytokines, phagocytose tumor cells, and present antigens to T cells. Mechanistically, Carfilzomib elicited unfolded protein response (UPR), activated IRE1α to recruit TRAF2, and activated NF-κB to transcribe genes encoding M1 markers in M2 macrophages. In vivo, Carfilzomib effectively rewired tumor microenvironment through reprogramming TAMs into M1-like macrophages and shrank autochthonous lung cancers in transgenic mouse model. More importantly, Carfilzomib synergized with PD-1 antibody to almost completely regress autochthonous lung cancers. Given the safety profiles of Carfilzomib in clinic, our work suggested a potentially immediate application of combinational treatment with Carfilzomib and PD-1 inhibitors for patients with solid tumors.
Sigma-Aldrich
Ovalbumin (323-339) (chicken, Japanese quail)
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
Diphtheria Toxin from Corynebacterium diphtheriae, lyophilized powder, Product is in unnicked form