Skip to Content
Merck
All Photos(1)

Key Documents

O1641

Sigma-Aldrich

Ovalbumin (323-339) (chicken, Japanese quail)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

Assay

97-100% (HPLC)

Quality Level

form

lyophilized powder

composition

peptide content, 65-100%

color

white

solubility

water, double-distilled: soluble

storage temp.

−20°C

Amino Acid Sequence

H-Ile-Ser-Gln-Ala-Val-His-Ala-Ala-His-Ala-Glu-Ile-Asn-Glu-Ala-Gly-Arg-OH

General description

Ovalbumin (323-339) is an egg white protein surrounded by B-cell epitopes, which are bound by specific IgE antibodies. In addition to this, it also contains CD4+ T cell epitopes.
The peptide is a major MHC class II-restricted T-cell epitope of Ovalbumin which is used for studies of immunological allergic responses such as asthma, food allergy and more..

Application

Ovalbumin (323-339) (chicken, Japanese quail) has been used to induce allergic asthma in mice.

Biochem/physiol Actions

Ovalbumin (OVA) peptide 323-339 mediates 25–35% of T-cell response to intact OVA in isolated BALB/c mouse. OVA 323-339 has been widely used to determine the characteristics of class II major histocompatibility complex (MHC)-peptide binding and T-cell activation.
With the help of two functionally distinct registers ovalbumin (323-339) peptide can bind to histocompatibility complex class II I-A(d) protein.

Physical form

Lyophilized from 0.1% aqueous TFA

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Zhou et al.
EMBO molecular medicine, 14(1), e14502-e14502 (2021-12-14)
Impressive clinical benefit is seen in clinic with PD-1 inhibitors on portion of cancer patients. Yet, there remains an urgent need to develop effective synergizers to expand their clinical application. Tumor-associated macrophage (TAM), a type of M2-polarized macrophage, eliminates or
Weifang Wang et al.
Frontiers in nutrition, 9, 931427-931427 (2022-08-02)
Asthma is a chronic disease, which is harmful to the health of the body and the quality of life. Supplementation of Lactobacillus can affect the immune environment of the lungs through the gut-lung axis. This study aimed to explore the
Haruyo Nakajima-Adachi et al.
Bioscience, biotechnology, and biochemistry, 76(10), 1979-1981 (2012-10-11)
The epitopes for OVA323-339-specific CD4⁺T cells from OVA23-3 food allergy model and DO11.10 tolerant induction model mice were analyzed. We found that OVA23-3 CD4⁺T cells recognized the N-terminal region, showing strong proliferation and the Th2-phenotype, and that DO11.10 CD4⁺T cells
Min Liu et al.
Immunity, inflammation and disease, 11(2), e730-e730 (2023-02-18)
Asthma is a chronic respiratory disease with an increasing incidence every year. microRNAs (miRNAs) have been demonstrated to have implications for asthma. However, limited information is available regarding the effect of miR-124-3p on this disease. Therefore, this study aimed to
S Sung et al.
Journal of immunology (Baltimore, Md. : 1950), 166(2), 1261-1271 (2001-01-06)
Dendritic cells (DC) are the primary APC responsible for the capture of allergens in the airways and the shuttling of processed allergens to the draining lymph nodes where Ag presentation and T cell activation take place. The mechanism of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service