Merck
  • USP8 inhibition reshapes an inflamed tumor microenvironment that potentiates the immunotherapy.
USP8 inhibition reshapes an inflamed tumor microenvironment that potentiates the immunotherapy.
Nature communications (2022-04-02)
Wenjun Xiong, Xueliang Gao, Tiantian Zhang, Baishan Jiang, Ming-Ming Hu, Xia Bu, Yang Gao, Lin-Zhou Zhang, Bo-Lin Xiao, Chuan He, Yishuang Sun, Haiou Li, Jie Shi, Xiangling Xiao, Bolin Xiang, Conghua Xie, Gang Chen, Haojian Zhang, Wenyi Wei, Gordon J Freeman, Hong-Bing Shu, Haizhen Wang, Jinfang Zhang
ABSTRACT
Anti-PD-1/PD-L1 immunotherapy has achieved impressive therapeutic outcomes in patients with multiple cancer types. However, the underlined molecular mechanism(s) for moderate response rate (15-25%) or resistance to PD-1/PD-L1 blockade remains not completely understood. Here, we report that inhibiting the deubiquitinase, USP8, significantly enhances the efficacy of anti-PD-1/PD-L1 immunotherapy through reshaping an inflamed tumor microenvironment (TME). Mechanistically, USP8 inhibition increases PD-L1 protein abundance through elevating the TRAF6-mediated K63-linked ubiquitination of PD-L1 to antagonize K48-linked ubiquitination and degradation of PD-L1. In addition, USP8 inhibition also triggers innate immune response and MHC-I expression largely through activating the NF-κB signaling. Based on these mechanisms, USP8 inhibitor combination with PD-1/PD-L1 blockade significantly activates the infiltrated CD8+ T cells to suppress tumor growth and improves the survival benefit in several murine tumor models. Thus, our study reveals a potential combined therapeutic strategy to utilize a USP8 inhibitor and PD-1/PD-L1 blockade for enhancing anti-tumor efficacy.
Product Description
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
DUB-IN-2, ≥98% (HPLC)
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-CMTM6 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution