Skip to Content
Merck
All Photos(1)

Key Documents

A2095

Millipore

Monoclonal Anti-HA−Agarose antibody produced in mouse

clone HA-7, purified immunoglobulin, PBS suspension

Synonym(s):

Monoclonal Anti-HA, Anti-HA, Anti-Influenza Hemagglutinin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.56
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

HA-7, monoclonal

form

PBS suspension

isotype

IgG1

capacity

20-50 nmol/mL elution capacity (HA-tagged fusion protein)
30-50 nmol/mL binding capacity (HA-tagged fusion protein)

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

The HA tag is a 9-amino acid tag, with the sequence YPYDVPDYA (N-Tyr-Pro-Tyr-Asp-Val-Pro-Asp-Tyr-Ala-C), derived from the hemagglutinin (HA) protein at amino acid residues 98-106 in the HA sequence. The HA tag contains a high proportion of charged amino acid residues, which makes the HA tag likely to form a strong antibody recognition site. Monoclonal Anti-HA Agarose Conjugate recognizes native as well as denatured/reduced forms of HA-tagged proteins. This product is reactive with N-or C-terminal HA-tagged fusion proteins that are expressed in E. coli or in mammalian cells.

Specificity

The antibody recognizes native as well as denatured-reduced forms of HA-tagged proteins and is reactive with N- or C-terminal HA-tagged fusion proteins expressed in E. coli or in mammalian cells.

Immunogen

synthetic peptide corresponding to a fragment of human influenza virus hemagglutinin (HA) known as HA-tag, conjugated to KLH

Application

Suitable for use in immunoprecipitation assays and for immunoaffinity purification of HA-tagged fusion proteins.

Physical form

Suspension in 0.01 M phosphate buffered saline, containing 15 mM sodium azide

Preparation Note

Prepared using cyanogen bromide-activated agarose. Specific antibody concentration is 2.0 - 2.4 mg/mL settled resin.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lin-Chun Chang et al.
Nucleic acids research, 40(3), 1331-1344 (2011-10-15)
The yeast RNA helicase Dhh1p has been shown to associate with components of mRNA decay and is involved in mRNA decapping and degradation. An RNA-binding protein, Rbp1p, is known to bind to the 3'-UTR of porin (POR1) mRNA, and induces
Frontiers in neurology and neuroscience research null
Ying Li et al.
Cancer research, 70(20), 8159-8168 (2010-10-05)
As a tumor marker for colorectal cancers, carcinoembryonic antigen (CEA) enhances the metastatic potential of cancer cells. CEA functions as an intercellular adhesion molecule and is upregulated in a wide variety of human cancers. However, the molecular mechanisms by which
Yinghong Ma et al.
Journal of virology, 85(1), 86-97 (2010-10-22)
Many aspects of the assembly of hepatitis C virus (HCV) remain incompletely understood. To characterize the role of NS2 in the production of infectious virus, we determined NS2 interaction partners among other HCV proteins during productive infection. Pulldown assays showed
Luzhou Xing et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 24(1-2), 30-43 (2012-10-26)
The chromosome 21 gene RCAN1, encoding a modulator of the calcineurin (CaN) phosphatase, is a candidate gene for contributing to cognitive disability in people with Down syndrome (DS; trisomy 21). To develop a physiologically relevant model for studying the biochemistry

Questions

1–2 of 2 Questions  
  1. What is the isotype and light chain for Monoclonal Anti-HA antibody clone HA-7 produced in mouse?

    1 answer
    1. Monoclonal Anti-HA antibody clone HA-7 is mouse IgG1, kappa light chain.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service