Merck
3-D-QSAR and docking studies on the neuronal choline transporter.
Bioorganic & medicinal chemistry letters (2010-07-20)
Werner J Geldenhuys, David D Allen, Paul R Lockman
ABSTRACT
The high affinity neuronal choline transporter (CHT1) is responsible for the uptake of choline into the pre-synaptic terminal of cholinergic neurons. Considering our past experience with modeling the blood-brain barrier choline transporter (BBBCHT) as drug delivery vector to the CNS, we investigated the 3-D-quantitative structure-activity relationship of the neuronal choline transporter. Comparative molecular field analysis (CoMFA) and comparative similarity index analysis (CoMSIA) yielded cross-validated models with a q(2) of 0.5, and a non-cross validated r(2) of 0.8. The electrostatic results of the 3-D-QSAR models are corroborated with a docking study into the bacterial choline transporter. Using this electrostatic map, we propose a putative binding site in a homology model of the CHT1. Knowledge gained from this study is useful to better understand the CHT1 as well as can be used in medicinal chemistry programs targeting this transporter.
Sigma-Aldrich
Trimethylamine, anhydrous, ≥99%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Dimethylamine solution, 40 wt. % in H2O
Sigma-Aldrich
3-Dimethylamino-1-propanol, 99%
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Sigma-Aldrich
3,4-Diaminopyridine, ≥98%
Sigma-Aldrich
2-Dimethylaminoethanol, ≥99.5%
Sigma-Aldrich
2-Dimethylaminoethanol, purified by redistillation, ≥99.5%
Sigma-Aldrich
(±)-Nicotine, ≥99% (TLC), liquid
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetramethylammonium chloride solution, for molecular biology
Sigma-Aldrich
2-(Methylamino)ethanol, ≥98%
Sigma-Aldrich
2-Dimethylaminoethanol, purum, ≥98.0% (GC)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
2,4,6-Trimethylpyridine, puriss. p.a., 99% (GC)
Sigma-Aldrich
3-Quinuclidinol, 99%
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in propylene glycol
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, ≥99%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC/NT)