Merck
  • Inhibitory effects of benzyl benzoate and its derivatives on angiotensin II-induced hypertension.
Inhibitory effects of benzyl benzoate and its derivatives on angiotensin II-induced hypertension.
Bioorganic & medicinal chemistry (2008-08-02)
Osamu Ohno, Mao Ye, Tomoyuki Koyama, Kazunaga Yazawa, Emi Mura, Hiroshi Matsumoto, Takao Ichino, Kaoru Yamada, Kazuhiko Nakamura, Tomohiro Ohno, Kohji Yamaguchi, Junji Ishida, Akiyoshi Fukamizu, Daisuke Uemura
ABSTRACT
Hypertension is a lifestyle-related disease which often leads to serious conditions such as heart disease and cerebral hemorrhage. Angiotensin II (Ang II) plays an important role in regulating cardiovascular homeostasis. Consequently, antagonists that block the interaction of Ang II with its receptors are thought to be effective in the suppression of hypertension. In this study, we searched for plant compounds that had antagonist-like activity toward Ang II receptors. From among 435 plant samples, we found that EtOH extract from the resin of sweet gum Liquidambar styraciflua strongly inhibited Ang II signaling. We isolated benzyl benzoate and benzyl cinnamate from this extract and found that those compounds inhibited the function of Ang II in a dose-dependent manner without cytotoxicity. An in vivo study showed that benzyl benzoate significantly suppressed Ang II-induced hypertension in mice. In addition, we synthesized more than 40 derivatives of benzyl benzoate and found that the meta-methyl and 3-methylbenzyl 2'-nitrobenzoate derivatives showed about 10-fold higher activity than benzyl benzoate itself. Thus, benzyl benzoate, its derivatives, and benzyl cinnamate may be useful for reducing hypertension.
Sigma-Aldrich
Benzyl benzoate, SAJ first grade, ≥98.0%
Sigma-Aldrich
Benzyl benzoate, tested according to Ph. Eur.
Sigma-Aldrich
Benzyl cinnamate, 99%
Sigma-Aldrich
Benzyl benzoate, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl cinnamate, ≥98%, stabilized
Sigma-Aldrich
Benzyl benzoate, natural, ≥99%, FG
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Supelco
Benzyl benzoate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Benzyl benzoate, analytical standard
Sigma-Aldrich
Benzyl benzoate, meets USP testing specifications