Skip to Content
Merck
All Photos(2)

Documents

68183

Supelco

Benzyl benzoate

analytical standard

Synonym(s):

Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COOCH2C6H5
CAS Number:
Molecular Weight:
212.24
Beilstein:
2049280
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

1 mmHg ( 125 °C)

Assay

≥98% (GC)

autoignition temp.

896 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.568 (lit.)

bp

323-324 °C (lit.)

mp

17-20 °C (lit.)

density

1.118 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl benzoate, an ester of benzyl alcohol and benzoic acid, is widely used as a fragrance fixer, fragrance ingredient and preservative to maintain the potency and stability of a variety of cosmetic formulations. It is also employed as a synthetic musk, acaricide to treat scabies and headlice.

Application

Benzyl benzoate may be used as an analytical reference standard for the quantification of the analyte in commercial products and pharmaceutical preparations using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of preservatives in cosmetic products: II. High-performance liquid chromatographic identification
Kruijf DN, et al.
Journal of Chromatography A, 469(5), 317-328 (1989)
Benzyl Benzoate-Loaded Microemulsion for Topical Applications: Enhanced Dermatokinetic Profile and Better Delivery Promises
Sharma G, et al.
Aaps Pharmscitech, 17(5), 1221-1231 (2016)
Photochemical products of benzyl benzoate: possible formation of skin allergens
Shibamoto T and Umano K
Cutaneous and Ocular Toxicology, 4(2), 97-103 (1985)
Separation and simultaneous high-performance liquid chromatographic determination of benzocaine and benzyl benzoate in a pharmaceutical preparation
Gigante B, et al.
Journal of Chromatography A, 549(2-3), 217-220 (1991)
Simultaneous micro steam distillation/solvent extraction for the isolation of semivolatile flavor compounds from cinnamon and their separation by series coupled-column gas chromatography
Jayatilaka A, et al.
Analytica Chimica Acta, 302(2-3) (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service